Skip to Content
Merck
All Photos(1)

Key Documents

SYMBXTC01

Millipore

Steritest® Symbio connection cable extension

With tri-clover clamp, for use with Steritest® Symbio LFH or FLEX

Synonym(s):

Pump connection wire

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
€1,430.00

€1,430.00


Available to ship on15 April 2025Details


Request more information

Select a Size

Change View
1 UNIT
€1,430.00

About This Item

UNSPSC Code:
23151818
NACRES:
NB.79

€1,430.00


Available to ship on15 April 2025Details


Request more information

material

PVC cable(s) (Connection)

Agency

EP (2.6.1)
JP (4.06)
USP (71)

packaging

pkg of 1 kit

manufacturer/tradename

Steritest®

input

pharmaceutical(s)

application(s)

pharmaceutical
sterility testing

compatibility

for use with Steritest® Symbio FLEX Pump Kit, 2 media (SYMBFLE01)
for use with Steritest® Symbio LFH Pump Kit (SYMBLFH01)
for use with Steritest® Symbio LFH or FLEX

shipped in

ambient

General description

Steritest® Symbio connection cable extension with Tri-Clover clamp is an accessory of Steritest® Symbio LFH and FLEX Pumps.

Application

Steritest® Symbio connection cable extension with tri-clover clamp is used to connect the Steritest® Symbio LFH or FLEX Pump to the communication hub when placed on an isolator table without hole.

Legal Information

STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service