Skip to Content
Merck
All Photos(5)

Key Documents

1.09626

Supelco

Benzyl alcohol

for analysis EMSURE®

Synonym(s):

Benzenemethanol

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€156.00
2.5 L
€324.00
25 L
€1,880.00

€156.00


Estimated to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
1 L
€156.00
2.5 L
€324.00
25 L
€1,880.00

About This Item

Linear Formula:
C6H5CH2OH
CAS Number:
Molecular Weight:
108.14
Beilstein:
878307
MDL number:
UNSPSC Code:
12191501
EC Index Number:
202-859-9
NACRES:
NB.21
grade:
reagent grade
bp:
203-205 °C (lit.)
vapor pressure:
0.07 hPa ( 20 °C)
13.3 mmHg ( 100 °C)
3.75 mmHg ( 77 °C)

€156.00


Estimated to ship on31 March 2025Details


Request a Bulk Order

grade

reagent grade

Quality Level

vapor density

3.7 (vs air)

vapor pressure

0.07 hPa ( 20 °C)
13.3 mmHg ( 100 °C)
3.75 mmHg ( 77 °C)

product line

EMSURE®

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

435 °C (DIN 51794)
817 °F

potency

1620 mg/kg LD50, oral (Rat)

dilution

(for analytical testing)

impurities

≤0.000002% B (Boron)
≤0.000002% Co (Cobalt)
≤0.000002% Cr (Chromium)
≤0.000002% Cu (Copper)
≤0.000002% Mn (Manganese)
≤0.000002% Ni (Nickel)
≤0.000005% Cd (Cadmium)
≤0.00001% Ba (Barium)
≤0.00001% Fe (Iron)
≤0.00001% Mg (Magnesium)
≤0.00001% Pb (Lead)
≤0.00001% Sn (Tin)
≤0.00001% Zn (Zinc)
≤0.00005% Al (Aluminium)
≤0.00005% Ca (Calcium)
≤0.001% Halogen compounds,halides (as Cl)
≤0.001 meq/g Acidity
≤0.002 meq/g Alkalinity
≤0.005% Sulfated ash
≤0.1% Benzaldehyde (GC)
≤0.1% Water

color

APHA: ≤10

refractive index

n20/D 1.539 (lit.)

bp

203-205 °C (lit.)

mp

−16-−13 °C (lit.)

peroxide value

≤5

transition temp

flash point 101 °C (DIN 51758)

solubility

40 g/L

SMILES string

OCc1ccccc1

density

1.045 g/mL at 25 °C (lit.)

storage temp.

2-30°C

InChI

1S/C7H8O/c8-6-7-4-2-1-3-5-7/h1-5,8H,6H2

InChI key

WVDDGKGOMKODPV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

EMSURE® grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE provides worldwide best and most extensive product specifications. We declare our EMSURE range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.
for analysis EMSURE®

Linkage

Discover EMSURE - to be more than sure!

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

213.8 °F - DIN 51758

Flash Point(C)

101 °C - DIN 51758


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service