Skip to Content
Merck
All Photos(1)

Key Documents

1.00801

Supelco

Label set acc. GHS

pkg of 20 labels blank adhesive labels (168 pictograms and 40 signal words), labels W × L 80 mm × 100 mm

Synonym(s):

Label set GHS

Sign Into View Organizational & Contract Pricing

Select a Size

1 SET
€104.00

€104.00


Estimated to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
1 SET
€104.00

About This Item

UNSPSC Code:
41122702
NACRES:
NB.51

€104.00


Estimated to ship on31 March 2025Details


Request a Bulk Order

packaging

pkg of 20 labels blank adhesive labels (168 pictograms and 40 signal words)

labels W × L

80 mm × 100 mm

storage temp.

room temp

General description

With the implementation of the CLP/EU-GHS (Global Harmonized System) directive in January 2009, the procedures for the classification, labeling, and packaging of hazardous chemicals have been redefined. Preparing mixtures, such as e.g. mobile phases for HPLC, is a routine part of the work in a lab. All chemical containers should be properly labeled in order to minimize the risk of confusing chemicals. This enables first responders to initiate the correct measures in the event of an accident.
with 20 blank adhesive labels 80 x 110 mm, 168 adhesive hazard pictograms, 40 adhesive signal words, package insert with H- and P-statements

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service