Skip to Content
Merck
All Photos(1)

Documents

1.08801

Supelco

Bottle key

for use with bottles with S 40 and S 28 screw caps

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122409
NACRES:
NB.51

compatibility

for use with bottles with S 40 and S 28 screw caps

storage temp.

no temp limit

General description

With this specially developed bottle key our S28 and S40 bottle screw caps can be opened easily to increase safety in daily solvents usage.
for opening and closing bottles with S 40 and S 28 screw caps

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service