Skip to Content
Merck
All Photos(1)

Documents

1.08804

Supelco

Container key

for use with containers with KS 60x6 screw cap

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122409
NACRES:
NB.51

compatibility

for use with containers with KS 60x6 screw cap

storage temp.

no temp limit

Related Categories

General description

for opening containers with KS 60x6 screw cap

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service