Skip to Content
Merck
All Photos(1)

Key Documents

1.07070

Supelco

Antistatic device

cable L 2 m

Synonym(s):

Grounding line

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€292.00

€292.00


Estimated to ship on26 March 2025Details


Request a Bulk Order

Select a Size

Change View
1 EA
€292.00

About This Item

UNSPSC Code:
41121815
NACRES:
NA.03

€292.00


Estimated to ship on26 March 2025Details


Request a Bulk Order

cable L

2 m

storage temp.

no temp limit

General description

If flammable liquids (e.g. solvents) are to be withdrawn or filled, the container must be properly grounded according to valid safety regulations to avoid explosions and fire risks. The relevant measures are to be taken to discharge static electricity. This can be done by using the antistatic device set. It contains 3 cables for highest flexibility in grounding containers during all work steps. It comes in a suitcase for safe and professional storage of cables that are currently not in use. Cable length: 2 m
for earthing metal containers when dispensing and filling flammable solvents

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service