1.01781
Petroleum benzine boiling range 100-120°C
for analysis EMSURE® Reag. Ph Eur
Synonym(s):
Petroleum ether, Petroleum spirit, Petroleum ether
About This Item
Recommended Products
grade
reagent grade
Quality Level
Agency
reag. Ph. Eur.
vapor pressure
30-40 hPa ( 20 °C)
product line
EMSURE®
form
liquid
autoignition temp.
250 °C
potency
>5000 mg/kg LD50, oral (Rat)
expl. lim.
0.8-6.5 % (v/v)
dilution
(for analytical testing)
impurities
≤0.000002% B (Boron)
≤0.000002% Co (Cobalt)
≤0.000002% Cr (Chromium)
≤0.000005% Cd (Cadmium)
≤0.00001% Ba (Barium)
≤0.00005% Al (Aluminium)
≤0.00005% Ca (Calcium)
≤0.0003 meq/g Acidity
≤0.002% Aromatics (as C6H6)
≤0.005% Sulfur compounds (as S)
≤0.01% Water
evapn. residue
≤0.001%
kinematic viscosity
0.6 cSt(20 °C)
bp
100-125 °C/1013 hPa
iodine value
≤0.3
transition temp
flash point <-10 °C
density
0.70 g/cm3 at 20 °C
cation traces
Al: ≤0.00005%
B: ≤0.000002%
Ba: ≤0.00001%
Ca: ≤0.00005%
Cd: ≤0.000005%
Co: ≤0.000002%
Cr: ≤0.000002%
Cu: ≤0.000002%
Fe: ≤0.00001%
Mg: ≤0.00001%
Mn: ≤0.000002%
Ni: ≤0.000002%
Pb: ≤0.00001%
Sn: ≤0.00001%
Zn: ≤0.00001%
storage temp.
2-30°C
Looking for similar products? Visit Product Comparison Guide
General description
Linkage
Other Notes
Legal Information
accessory
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
14.0 °F
Flash Point(C)
-10 °C
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
EU REACH Annex XVII (Restriction List)
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service