Skip to Content
Merck
All Photos(1)

Documents

C-236

Supelco

Cannabinolic Acid-D3 (CBNA-D3) solution

100 μg/mL (1% DIPEA and 0.05% Ascorbic Acid in Acetonitrile), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H23D3O4
Molecular Weight:
357.46
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL (1% DIPEA and 0.05% Ascorbic Acid in Acetonitrile)

format

single component solution

storage temp.

−70°C

General description

This stable-labeled internal standard is suitable for quantitation of cannabinolic acid levels by LC-MS/MS and GC/MS applications for urine drug testing, pharmaceutical research, clinical toxicology, cannabis testing or forensic analysis. Cannabinolic Acid (CBNA) is a non-psychoactive cannabinoid found in Cannabis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service