Skip to Content
Merck
All Photos(1)

Documents

C-269

Supelco

Cannabinoid Mixture - 17 Components solution

new

250 μg/mL ( 0.1% ascorbic acid and 1% DIPEA in 50:50 MeOH:ACN, each analyte), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

composition

Cannabichromenic acid (CBCA), 250 μg/mL
Cannabicyclol (CBL), 250 μg/mL
Cannabicyclolic acid (CBLA), 250 μg/mL
Cannabidiol, 250 μg/mL
Cannabidivarinic acid (CBDVA), 250 μg/mL
Cannabigerol (CBG), 250 μg/mL
Cannabinol, 250 μg/mL
Cannabinolic acid (CBNA), 250 μg/mL
CBC (Cannabichromene), 250 μg/mL
CBDA (Cannabidiolic acid), 250 μg/mL
CBDV (Cannabidivarin), 250 μg/mL
CBGA (Cannabigerolic acid), 250 μg/mL
(-)-Delta8-THC, 250 μg/mL
(-)-Delta9-THC, 250 μg/mL
Tetrahydrocannabivarinic acid (THCVA), 250 μg/mL
THCA-A (delta-9-Tetrahydrocannabinolic acid A), 250 μg/mL
THCV (delta-9-Tetrahydrocannabivarin), 250 μg/mL

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

250 μg/mL ( 0.1% ascorbic acid and 1% DIPEA in 50:50 MeOH:ACN, each analyte)

format

multi-component solution

storage temp.

-65 to -96°C

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

41.0 °F - closed cup

Flash Point(C)

5 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service