Skip to Content
Merck
All Photos(1)

Key Documents

C-228

Supelco

Cannabidiol Hydroxyquinone (HU-331)

certified reference material, vial of 5 mg, Cerilliant®

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€443.00

€443.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€443.00

About This Item

Empirical Formula (Hill Notation):
C21H28O3
CAS Number:
Molecular Weight:
328.45
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€443.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

packaging

vial of 5 mg

manufacturer/tradename

Cerilliant®

format

neat

storage temp.

-65 to -96°C

InChI

1S/C21H28O3/c1-5-6-7-8-15-12-18(22)19(21(24)20(15)23)17-11-14(4)9-10-16(17)13(2)3/h11-12,16-17,24H,2,5-10H2,1,3-4H3/t16-,17+/m0/s1

InChI key

WDXXEUARVHTWQF-DLBZAZTESA-N

General description

Cannabidiol Hydroxyquinone, also known as HU-331, is an anticarcinogenic drug synthesized from cannabidiol (CBD) showing great efficacy against oncogenic human cells. HU-331 has also been identified to be a CBD degradant. This certified Snap-N-Spike® solution is suitable for cannabidiol hydroxyquinone testing methods by GC/MS, LC/MS or HPLC for applications in clinical toxicology, impurity profiling, pharmaceutical research, forensic analysis, and urine drug testing.
Snap-N-Spike®/Snap-N-Shoot®

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service