Skip to Content
Merck
All Photos(1)

Documents

C-143

Supelco

Cannabichromene solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

CBC solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30O2
CAS Number:
Molecular Weight:
314.46
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

cannabis testing
cannabis testing

format

single component solution

shipped in

wet ice

storage temp.

−20°C

InChI

1S/C21H30O2/c1-5-6-7-10-17-14-19(22)18-11-13-21(4,23-20(18)15-17)12-8-9-16(2)3/h9,11,13-15,22H,5-8,10,12H2,1-4H3

InChI key

UVOLYTDXHDXWJU-UHFFFAOYSA-N

General description

This Certified Snap-N-Spike® solution standard is suitable for cannabichromene testing methods by GC/MS, HPLC or LC-MS/MS for Cannabis potency testing or impurity profiling, pharmaceutical research, and forensic analysis. Cerilliant solution Certified Reference Materials (CRMs) of cannabinoids are supplied in a convenient, quantitative, US DEA-exempt solution format and with TK#s for Canadian customers.

Cannabichromene (CBC) is a non-psychoactive cannabinoid that belongs to the class of C21 terpenophenolic compounds and reportedly exerts anti-inflammatory, antimicrobial, and analgesic activity.

Application

The Certified Reference Material (CRM) in solution can also be used as follows:
  • Multi-analysis of 11 cannabinoids in biomass and extracts of different cannabis varieties by an HPLC-UV method, following the International Conference on Harmonization (ICH) Tripartite Guideline for Validation of Analytical Procedures
  • Analysis of dried marijuana flowers to measure the amount of nine cannabinoids by high-performance liquid chromatography (HPLC) along with UV detection
  • Development and validation of an ultra high-performance liquid chromatographic (UHPLC) method coupled to photodiode array and mass spectrometry detectors to determine 11 cannabinoids in various Cannabis Sativa samples
  • Simultaneous detection of seven cannabinoids from cannabis oil extract samples by gas chromatography-mass spectrometry (GC-MS)
  • Gas chromatographic-mass spectrometric (GC-MS) analysis of ten main cannabinoids from hemp inflorescences following their silylation and esterification for derivatization

Features and Benefits

  • Fully characterized under ISO/IEC 17025 and ISO 17034 accreditation
  • Accompanied with a comprehensive Certificate of Analysis (CoA) with data on stability, homogeneity, accuracy of concentration, uncertainty, and traceability
  • Rigorously tested through real-time stability studies to ensure accuracy and shelf life
  • Gravimetrically prepared using qualified precision balances to ensure minimal uncertainty
  • Flame sealed under argon into ampoules for long-term shelf life
  • Offered in a convenient, DEA-exempt format to improve laboratory efficiency

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Cannabis products need cannabinoid characterization. Ascentis Express C18 resolves 14 similar cannabinoids in under seven minutes with excellent peak shape.

ISO-certified cannabinoid CRMs, THC and THCA, available as single components or mixes, for accurate potency testing.

HPLC-DAD workflow analyzes 14 cannabinoids in hemp extracts in 10 minutes using Chromolith columns for robust performance.

A complete workflow for the HPLC separation of 17 cannabinoids in different cannabis products using Ascentis® Express C18 Column in a shorter run time with retention time stability. Learn more here.

See All

Protocols

Cannabis products need cannabinoid characterization. Ascentis Express C18 resolves 14 similar cannabinoids in under seven minutes with excellent peak shape.

HPLC separation of 17 important cannabinoids including CBD, delta 9 THC and THCA. Read the application note.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service