Skip to Content
Merck
All Photos(1)

Key Documents

57021

Supelco

Empty SPE Tube (no frits)

volume 20 mL, pk of 12

Synonym(s):

SPE Reservoir

Sign Into View Organizational & Contract Pricing

Select a Size

12 EA
$85.30

$85.30


Check Cart for Availability

Request a Bulk OrderRequest a Sample

Select a Size

Change View
12 EA
$85.30

About This Item

UNSPSC Code:
12352200

$85.30


Check Cart for Availability

Request a Bulk OrderRequest a Sample

material

polypropylene

packaging

pk of 12

technique(s)

solid phase extraction (SPE): suitable

volume

20 mL

application(s)

food and beverages

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

When used in conjunction with SPE Tube Adapters, empty SPE tubes can be stacked on top of a standard SPE cartridge during processing. The empty SPE cartridge acts as a reseroir to accommodate larger sampling volumes.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service