Skip to Content
Merck
All Photos(1)

Key Documents

57242

Supelco

Empty SPE Tube (no frits)

volume 6 mL, pkg of 30 ea

Synonym(s):

SPE Reservoir

Sign Into View Organizational & Contract Pricing

Select a Size

30 EA
$30.00

$30.00


Available to ship on14 April 2025Details


Request a Bulk OrderRequest a Sample

Select a Size

Change View
30 EA
$30.00

About This Item

UNSPSC Code:
12000000

$30.00


Available to ship on14 April 2025Details


Request a Bulk OrderRequest a Sample

material

polypropylene

packaging

pkg of 30 ea

technique(s)

solid phase extraction (SPE): suitable

volume

6 mL

application(s)

environmental
food and beverages

compatibility

for use with S10L Elution reservoir

Looking for similar products? Visit Product Comparison Guide

General description

Solid phase extraction apparatus uses glass vials or tubes for sample collection and concentration. Some sorbents require large desorption volume than small SPE cartridges in such case SPE tubes were used.[1]

Application

Empty SPE Tube (no frits) was coupled with closed-face polystyrene cassette containing glass fibre filters in a method to reduce the interference from vapour solvent caused in vapour fraction, during mist sampling and analysis, in an experiment conducted to study the occupational exposure to mineral oil metalworking fluid (MWFs) Mist.[2]

Features and Benefits

When used in conjunction with SPE Tube Adapters, empty SPE tubes can be stacked on top of a standard SPE cartridge during processing. The empty SPE cartridge acts as a reseroir to accommodate larger sampling volumes.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Occupational exposure to mineral oil metalworking fluid (MWFs) mist: development of new methodologies for mist sampling and analysis. Results from an inter-laboratory comparison.
Huynh, C. Khanh, et al.
Journal of Physics. Conference Series, 151 (2009)
Nigel J.K. Simpson
Solid-Phase Extraction: Principles, Techniques, and Applications, 445-445 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service