Key Documents
51272-U
MicroTight Adapters
PEEK, for use with (1/16" to 1/32" tubing)
Select a Size
1 EA
$81.40
$81.40
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
$81.40
About This Item
UNSPSC Code:
41115700
NACRES:
SB.52
$81.40
Check Cart for Availability
Request a Bulk Order
material
PEEK
compatibility
for use with (1/16" to 1/32" tubing)
General description
These adapters will help create a true zero dead volume (ZDV) connection between 1/16" I.D. tubing and capillary tubing. MicroTight Adapters allow connection of 1/16" O.D. tubing directly to 1/32" O.D. and 360 μm tubing.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Choose from one of the most recent versions:
Certificates of Analysis (COA)