Skip to Content
Merck
All Photos(2)

Key Documents

55067-U

Upchurch PEEK Fingertight Fitting

10-32 fitting (one-piece), pkg of 10 ea

Synonym(s):

Upchurch PEEK Finger-tight Fitting

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
$220.00

$220.00


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
10 EA
$220.00

About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

$220.00


Available to ship on22 April 2025Details


Request a Bulk Order

description

fitting total L 13/16 in. head width 3/8 in.

packaging

pkg of 10 ea

I.D.

0.0625 in. (1.5875 mm)

fitting

10-32 fitting (one-piece)

Looking for similar products? Visit Product Comparison Guide

General description

The one-piece fitting, 55067-U, is convenient to use because the ferrule will not stick in a receiving port and can be easliy found if dropped onto the floor. The two-piece fitting features a separate ferrule so you can replace just the ferrule instead of the entire unit. The Upchurch PEEK Fingertight fittings are for use with 1/16″ O.D. tubing and can be used to 6,000 psi (420 kg/cm2).

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service