Skip to Content
Merck
All Photos(1)

Key Documents

501395

Supelco

Model 24 Single-Stage Regulator

for use with CGA-165 (for SCOTTY 48 cylinder)

Synonym(s):

MODEL 24 SCOTTY® IV REGUL. & GAUGE (CGA 165)

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$432.00

$432.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$432.00

About This Item

UNSPSC Code:
41112500

$432.00


Check Cart for Availability

Request a Bulk Order

description

single-stage

manufacturer/tradename

16-05-124SP165
Scott Gas

compatibility

for use with CGA-165 (for SCOTTY 48 cylinder)

Looking for similar products? Visit Product Comparison Guide

General description

Regulators are used to control volume of gas delivered in chromatography. There are mainly 2 types of regulators: single-stage and double-stage. Single-stage regulator shows slight variation in pressure delivery when cylinder pressure drops and also when flow rate increases. It also shows high ‘lockup′ pressure.[1]

Legal Information

SCOTTY is a registered trademark of Air Liquide

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Compressed Gas Association
Handbook of Compressed Gases, 48-48 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service