Key Documents
23884
In-Line Pressure Regulator
Ultra-High Purity version (outlet pressure 0-100 psi, 1/8 in. stainless steel fittings)
Select a Size
1 EA
$721.00
$721.00
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
$721.00
About This Item
UNSPSC Code:
41112500
$721.00
Check Cart for Availability
Request a Bulk Order
compatibility
Air, H2, N2, Ar, He
General description
Install an in-line pressure regulator downstream of a gas cylinder regulator to adjust pressure closer to the point of use. 400 psig maximum input pressure. 1/8 in. Swagelok nuts and ferrules included.
Application
Air, H2, N2, Ar, He
Choose from one of the most recent versions:
Certificates of Analysis (COA)