Key Documents
23315
Supelco
Flash Arrestor
Select a Size
1 EA
$559.00
$559.00
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
$559.00
About This Item
UNSPSC Code:
41112500
$559.00
Check Cart for Availability
Request a Bulk Order
General description
Install a flash arrestor downstream of a hydrogen gas cylinder (see figure), or a hydrogen generator. In event of a flashback, the flash arrestor diverts the flame into three feet (1 m) of tubing, where the flame is extinguished and the heat is absorbed. The shock wave preceding the flashback closes and locks the arrestor′s shutoff valve, eliminating the flow of gas. Inlet/outlet fittings are 1/4 in. female NPT. For use to 50 psig (3.5 kg/cm2). Meets Occupational Safety and Health Administration (OSHA) and National Fire Protection Agency (NFPA) codes. Factory Mutual approved.
Choose from one of the most recent versions:
Certificates of Analysis (COA)