Skip to Content
Merck
All Photos(2)

Key Documents

ACS500024

Millipore

Amicon® Pro Purification System

sample volume 10 mL, polypropylene tube (for collection)

Synonym(s):

Amicon Ultra-0.5 adapter

Sign Into View Organizational & Contract Pricing

Select a Size

24 EA
$307.00

$307.00


In StockDetails



Select a Size

Change View
24 EA
$307.00

About This Item

UNSPSC Code:
41104916
eCl@ss:
36100101
NACRES:
NB.24

$307.00


In StockDetails


material

polyethylene cap
polyethylene frit
polypropylene device (for exchange)
polypropylene frit
polypropylene tube (for collection)
polypropylene tube (holder)

Quality Level

sterility

non-sterile

product line

Amicon®

manufacturer/tradename

Amicon® Pro

parameter

10 mL sample volume

technique(s)

buffer exchange: suitable
protein purification: suitable (antibody labeling)
protein purification: suitable (concentration)
protein purification: suitable (enrichment)

L

12 cm (4.7 in.)

diam.

2.97 cm

shipped in

ambient

General description

The Amicon® Pro Purification System 24 pack includes 24 Exchange devices and 24 Collection tubes with cap.

Features and Benefits

Minimum Final Concentrate Volume: 15 µL

Physical form

Color Code: Clear with pink cap

Legal Information

Amicon is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Commercially available protein labeling kits provide researchers with the flexibility to customize their own immunoassay detection panels.

Protein purification is simpler with Amicon Pro. The device delivers consistent sample preparation for more reliable recovery, uncompromised purity and easier data generation.

Protocols

If an antibody must be purified from a small sample volume (0.5-10 mL), then the Amicon® Pro Purification System is recommended.

Detergents are frequently used to solubilize proteins and nucleic acids during purification, but the presence of detergents may interfere with downstream analyses.

Questions

  1. Has the product ACK5050NT been discontinued?

    1 answer
    1. The product ACK5050NT has been discontinued.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service