Skip to Content
Merck
All Photos(1)

Key Documents

20620

Supelco

Gas Line Filter

for use with 1/8 in. tubing

Synonym(s):

LINE FILTERS 1/8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41112500
Pricing and availability is not currently available.

material

brass (with replaceable stainless steel filter 25810-U)

compatibility

for use with 1/8 in. tubing

Looking for similar products? Visit Product Comparison Guide

General description

Pressure from gas line acts on a large area piston which drives a piston of small area. The pressure is amplified in the ratio of the areas of the faces of the pistons, and high pressure liquid at a constant pressure is delivered to the system. Line filter helps in constant pressure and drop in volume flow rate through the system.[1]
This brass gas line filter contains a replaceable 7 μm stainless steel filter, and will remove dust and small particles that could otherwise foul downstream needle valves. Nuts and ferrules included.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew Pryde, M.T. Gilbert
Applications of High Performance Liquid Chromatography, 14-14 (1979)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service