Key Documents
22789
Supelco
Fritted Purge Sampler
size 25 mL, Tekmar® 14-2334-024
Select a Size
1 EA
$290.00
$290.00
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
$290.00
About This Item
UNSPSC Code:
41115700
$290.00
Check Cart for Availability
Request a Bulk Order
material
glass
manufacturer/tradename
Tekmar® 14-2334-024
size
25 mL
application(s)
industrial hygiene
compatibility
for use with Tekmar® 2000/3000
for use with standard aqueous analysis
Features and Benefits
Note: All 1/4 in. O.D. glass ends are ground glass.
Legal Information
Tekmar is a registered trademark of Tekmar Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. Good morning, I would like to know if this glassware is compatible with Eclipse 4660 purge-and-trap O I analytical
    1 answer
    1. Fritted Purge Sampler, product 22789, is for use with Tekmar 2000/3000 and is also compatible with Atomx XYZ, Lumin, Stratum and Velocity.
      Helpful?