Key Documents
22749
Supelco
Fritted Purge Sampler
size 25 mL, top stem, Tekmar® 14-4007-024
Select a Size
1 EA
$335.00
$335.00
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
$335.00
About This Item
UNSPSC Code:
41115700
$335.00
Check Cart for Availability
Request a Bulk Order
material
glass
Agency
EPA 603
manufacturer/tradename
Tekmar® 14-4007-024
size
25 mL
application(s)
industrial hygiene
Features and Benefits
Note: All 1/4 in. O.D. glass ends are ground glass.
Legal Information
Tekmar is a registered trademark of Tekmar Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)