Key Documents
22003
Swagelok® Nut, Front and Back Ferrule Set
Swagelok® 400-S, brass, 1/4 in. Swagelok
Select a Size
10 EA
$50.10
$50.10
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
10 EA
$50.10
About This Item
UNSPSC Code:
41115717
$50.10
Check Cart for Availability
Request a Bulk Order
material
brass
packaging
pkg of 10 ea
manufacturer/tradename
Swagelok® 400-S
fitting
1/4 in. Swagelok
General description
Swage tubing coupler has four parts namely the body, into which the tube is placed, two ferrules that is front ferrule and back ferrule and lastly a packing nut.[1] While assembling a compression tube fitting, the metal tubing should be inspected properly to ensure that the surface is smooth. A nut is slided on to the tube fitting with the open side facing the end of the tubing. Followed by sliding the back ferrule with the wider part facing the nut. Further, sliding on the front ferrule with the small end of the cone facing the end of the tubing. Then inserting the tubing into the fitting and tighten up the nut/ferrule assembly onto the fitting.[2]
Application
Swagelok® nut, front and back ferrule set maybe used for sampler construction from a single capillary gas chromatographic column, for collection of semi-volatile organic compounds through a diffusion denuder.[3]
Legal Information
Swagelok is a registered trademark of Swagelok Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Diffusion denuder for the collection of semivolatile organic compounds.
Krieger MS and Hites RA.
Environmental Science & Technology, 26 (8), 1551-1555 (1992)
Grob RL & Barry EF.
Modern Practice of Gas Chromatography, 522-523 (2004)
The Manipulation of Air-Sensitive Compounds.
Shriver DF.
Nature, 217-218 (1986)