Key Documents
Z420085
STAT-O-LOK quick-disconnect
male, for tubing i.d., 0.25
Select a Size
1 EA
$132.00
$132.00
Check Cart for Availability
Select a Size
Change View
1 EA
$132.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
$132.00
Check Cart for Availability
feature
for tubing i.d. 0.25
manufacturer/tradename
Bel-Art
General description
  • Chrome-plated brass connectors with slide locks are leak-free
  • Automatic shut-off when disconnected
  • Hose barb ends accept flexible tubing
  • Operate at temperatures to 82°C (180°F), pressures to 250psi, vacuum to 30μm
  • Delrin® valves provide dependable operation
  • Ideal for liquid and gas connectors in biomedical instrumentation and general industrial applications
Legal Information
Delrin is a registered trademark of E. I. du Pont de Nemours and Company
STAT-O-LOK is a trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)