Key Documents
22039
Swagelok® Back Ferrule
Swagelok® 204-1, stainless steel, 1/8 in. Swagelok
Synonym(s):
Swagelok® 204-1,SS BACK FERRULE 1/8" PK5
Select a Size
5 EA
$34.40
$34.40
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
5 EA
$34.40
About This Item
UNSPSC Code:
41115717
$34.40
Check Cart for Availability
Request a Bulk Order
material
stainless steel
packaging
pkg of 5 ea
manufacturer/tradename
Swagelok® 204-1
fitting
1/8 in. Swagelok
General description
Swage type fittings are one of the most popular connectors which are available in metal and plastic forms; they are very useful for connecting similar or dissimilar materials, like metal to glass or metal to plastic. The general swage tubing coupler has four parts namely the body, into which the tube is placed, two ferrules that is front ferrule and back ferrule and lastly a packing nut.[1]
Legal Information
Swagelok is a registered trademark of Swagelok Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
The Manipulation of Air-Sensitive Compounds.
Shriver DF.
Nature, 217-218 (1986)