Skip to Content
Merck
All Photos(1)

Key Documents

CLS3985

Corning® 384 well microplate

Tissue Culture (TC)-treated surface, black polystyrene, square, sterile, lid, case of 100

Synonym(s):

384 multiwell plates, 384 well low volume plates, 384 well microplates, 384 well microtiter plates, 384 well plates, 384 well tissue culture treated plates

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
$1,160.00

$1,160.00


Available to ship on15 April 2025Details



Select a Size

Change View
100 EA
$1,160.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.15
material:
black polystyrene
clear bottom
flat wells
square
size:
384 wells

$1,160.00


Available to ship on15 April 2025Details


material

black polystyrene
clear bottom
flat wells
square

description

black, 384-well

sterility

sterile

feature

lid
skirt

packaging

case of 100

manufacturer/tradename

Corning 3985

size

384 wells

well volume

112 μL

well working volume

20-80 μL

wells

384 wells

suitability

suitable for (cell culture)

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

For optical imaging

Features and Benefits

  • Corning® 384 well microplate is ideal for whole cell or subcellular high content screening.
  • Black Polystyrene, TC-treated Microplates with flat clear bottom
  • Applicable for optical imaging
  • Suitable with high throughput confocal imagers
  • Optimized with the Amersham InCell 3000 Analyzer
  • Supports better cell growth and attachment than glass bottom plates
  • Black opaque walls minimize well-to-well crosstalk

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service