Skip to Content
Merck
All Photos(1)

Key Documents

CLS3825

Corning® 384 well plates

white polystyrene, flat bottom, non-sterile, lid: no, pack of 10

Synonym(s):

384 multiwell plates, 384 well microplates, 384 well microtiter plates, 384 well plates

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
$295.00
50 EA
$1,320.00

$295.00


Available to ship on22 April 2025Details



Select a Size

Change View
10 EA
$295.00
50 EA
$1,320.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
clear wells
flat bottom
white bottom
white polystyrene
size:
384 wells
sterility:
non-sterile
binding type:
non-treated surface
feature:
lid: no
skirt

$295.00


Available to ship on22 April 2025Details


material

clear wells
flat bottom
white bottom
white polystyrene

sterility

non-sterile

feature

lid: no
skirt

packaging

pack of 10
case of 50 ea

manufacturer/tradename

Corning 3825

size

384 wells

well volume

50 μL

color

white

binding type

non-treated surface

General description

  • Corning® 384 well plates, Low Volume 384-well White Flat Bottom Polystyrene is applicable for homogenous luminescent and homogeneous time resolved fluorescenc (HTRF) assays.
  • Not Treated Microplate without lid and not sterile
  • Designed to reduce well-to-well cross-talk during fluorescent and luminescent assays

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service