Skip to Content
Merck
All Photos(1)

Key Documents

P9116

Nunc® MicroWell 384 well optical bottom plates

384 well optical bottom plate, cell cultured treated, 120 μL/well, white, 30/cs

Synonym(s):

384 multiwell plate, 384 well ELISA plate, 384 well immunoassay plate, 384 well microplate, ELISA plate, optical bottom plate

Sign Into View Organizational & Contract Pricing

Select a Size

30 EA
$877.00

$877.00


Available to ship on07 April 2025Details



Select a Size

Change View
30 EA
$877.00

About This Item

UNSPSC Code:
41122107
NACRES:
NB.22
material:
flat bottom clear polystyrene wells
white polystyrene
size:
384 wells
sterility:
sterile
feature:
lid
polystyrene/polymer base

$877.00


Available to ship on07 April 2025Details


material

flat bottom clear polystyrene wells
white polystyrene

sterility

sterile

feature

lid
polystyrene/polymer base

packaging

pack of 10
case of 30 ea (internal packs of 10)

manufacturer/tradename

Nunc 142762

maximum volume

500 μL

plate L × W

128 mm × 86 mm

size

384 wells

well maximum volume

100 μm

working volume

100 μL

color

white

suitability

suitable for (fluorescence/luminescence based assays)

Looking for similar products? Visit Product Comparison Guide

General description

Polystyrene/Polymer Base

Optimum clarity for viewing well contents. Polymer bottom plates combine the optical clarity of virgin crystalline polystyrene with optimal surface for a wide range of HTS applications. Polystyrene upper structures available in black or white. Flat bottom well geometry for plate reader access. Black plates provide minimum back-scatterd light. White plates maximize reflection and provide minimum autoflorescence and autoluminescence. Note: All plates supplied with lid and sterile.

White or black upper structure with polystyrene/polymer bottom plates combine the optical clarity of virgin crystalline polystyrene with optimal surface for a wide range of HTS applications.

  • White or black upper structures with optically clear bottoms
  • Can be read from bottom and top, or both
  • Optical film made of polystyrene
  • Rounded square wells eliminate wicking (capillary action)
  • Optimized for scintillation counting
  • Nunclon® Δ (cell culture surface), Poly-D-Lysine or Collagen I surfaces for cell culture
  • Produced in clean environment
  • Working volume range: 10-100 μl/well
  • For research use only

Legal Information

MicroWell is a trademark of Thermo Fisher Scientific or its subsidiaries
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service