Skip to Content
Merck
All Photos(1)

Documents

CLS3742

Corning® Thermowell GOLD PCR 1 x 8 cap strips

Synonym(s):

1 x 8 cap strips, 1 x 8 optically clear flat caps, 8 well cap strips, 8 well optically clear flat caps

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.17

material

polypropylene

sterility

non-sterile

feature

autoclavable
designed to fit on all Thermowell GOLD 1 x 8 well PCR strips and 96 well PCR microplates
designed with an optically clear flat cap for RT-PCR and other fluorescent or luminescent applications

packaging

pack of 125
case of 1250

manufacturer/tradename

Corning 3742

technique(s)

PCR: suitable

color

clear

suitability

suitable for (PCR applications)

Looking for similar products? Visit Product Comparison Guide

General description

  • Made of thin wall polypropylene and designed to fit on all Thermowell GOLD 1 x 8 well PCR strips and 96 well PCR microplates
  • Certified DNase- and RNase-free
  • Autoclavable at 121°C

Application

For RT-PCR

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service