Skip to Content
Merck
All Photos(1)

Documents

11667009001

Roche

Strip PCR Tubes and Caps

pkg of 125 strips, tube volume <200 μL

Synonym(s):

PCR tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300

packaging

pkg of 125 strips

manufacturer/tradename

Roche

tube volume

<200 μL

shipped in

ambient

storage temp.

20-25°C

General description

The product contains thin-walled PCR tubes, attached to each other in convenient strips and corresponding strips of caps, which are not attached to the tube bodies. Each tube has a maximum volume of 200μl.

Application

Strip PCR Tubes and Caps are designed for use in PCR thermal cyclers, and provide optimal and rapid heat transfer. These tubes are ideally suited for use with the Expand PCR System, or with Titan One Tube RT-PCR in such applications as:
  • PCR
  • RT-PCR
  • cDNA synthesis
  • Sequencing/cycle sequencing

Quality

Absence of nucleases: Strips are DNase- and RNase-free according to the current Quality procedures.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service