Skip to Content
Merck
All Photos(1)

Key Documents

BR781414

BRAND® PCR caps, strips of 8

clear, domed polypropylene

Sign Into View Organizational & Contract Pricing

Select a Size

300 EA
$265.00

$265.00


Check Cart for Availability


Select a Size

Change View
300 EA
$265.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.22

$265.00


Check Cart for Availability

material

domed polypropylene

feature

RNase and DNase free

packaging

pkg of 25 × 12 strips (300 total)

manufacturer/tradename

BRAND 781414

technique(s)

PCR: suitable

color

clear

Looking for similar products? Visit Product Comparison Guide

General description

With PCR cap strips you can seal PCR strips and plates with 24, 48, and 96 wells reliably and protect your samples effectively against evaporation and contamination.The shape of the cap strips is optimally matched to BRAND PCR tube strips and plates so they are easy to open and close. To prevent contamination, the cap strips are equipped with two side tabs for exact positioning before closing and a small opening attachment on each cap. This prevents contact with the inside of the cap.The highly transparent flat cap strips are ideal for qPCR because the cap shape allows accurate fluorescence measurement. With domed cap strips, thermal cyclers without pressure sensors exert a stronger force, thus further increasing the sealing effect.BIO-CERT® PCR QUALITY: Free of human DNA, RNase, DNase, Pyrogens and PCR-Inhibitors; manufactured in cleanroom conditions.

Features and Benefits

  • Effective protection against evaporation and contamination
  • The right cap shape for your application
  • Easy to open and close without risk of contamination

Legal Information

BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service