Key Documents
CLS3004
Corning® single use spatulas
Small spoon/spoon
Synonym(s):
Spatula for Lab Use
Select a Size
100 EA
$378.00
$378.00
Check Cart for Availability
Select a Size
Change View
100 EA
$378.00
About This Item
UNSPSC Code:
41122403
NACRES:
NB.14
$378.00
Check Cart for Availability
sterility
sterile
manufacturer/tradename
Corning 3004
L
236 mm
General description
Corning single use spatulas have been designed to save time and provide contamination-free samples. Eliminates need to recycle and sterilize reusable spatulas.
Each spatula is individually packaged, RNase/DNase free, non-pyrogenic and antistatic.
Features and Benefits
  • Corning® single use spatulas have a narrow spoon at one end and a larger round spoon at the other end.
  • Polystyrene and color concentrate blend with 236 mm length.
  • Visually Inspected for accurate labeling, product configuration, seal and barrier integrity.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)