Z115282
PTFE-coated spatulas
Tapered to 1/8in., L 7 1/4 in.
All Photos(1)
About This Item
Recommended Products
packaging
pack of 2 ea
manufacturer/tradename
Bel-Art H36704-0000
L
7 1/4 in.
Related Categories
General description
2 per bag
Description: 4 mm diameter x 184 mm (0.156" x 7 1/4") tapered to .8 mm ( 1/32").
- Polished stainless steel tool with non stick Fluo-Kem® PTFE® FEP coating
- Double ended spatula, select blade style combination to match requirements
- FDA compliant for food contact
Description: 4 mm diameter x 184 mm (0.156" x 7 1/4") tapered to .8 mm ( 1/32").
Legal Information
Fluo-kem is a registered trademark of SP Scienceware, a division of SP
Teflon is a registered trademark of E. I. du Pont de Nemours and Company