Skip to Content
Merck
All Photos(1)

Key Documents

83462

Sigma-Aldrich

RBS 50 solution

concentrate

Synonym(s):

RBS Detergent 50

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161900
NACRES:
NA.21
Product 83462 is not currently sold in your country. Contact Technical Service

Quality Level

form

liquid

pH

11-12 (2% in H2O)

density

1.24 g/mL at 20 °C

General description

RBS 50 solution is a special detergent for glass and instruments in chemical, biological, and biotechnology laboratories. It is suitable for use in ultrasonic baths and automatic washing machines for laboratory glassware.

Application

RBS 50 solution has been used as a detergent to clean fluorine-doped tin oxide (FTO) glass substrates.[1]

Other Notes

Sales restrictions may apply

Legal Information

RBS is a trademark of Chemical Products R. Borghgraef S.A.

related product

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Enhanced performance of CH3NH3PbI3 perovskite solar cells via interface modification using phenyl ammonium iodide derivatives
Ghoreishi FS, et al.
Journal of Power Sources, 473, 228492-228492 (2020)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service