Skip to Content
Merck
All Photos(1)

Key Documents

30472

Sigma-Aldrich

DCN 90

concentrate

Synonym(s):

DCN Surfactant Detergent

Sign Into View Organizational & Contract Pricing

Select a Size

5 L
$438.00

$438.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
5 L
$438.00

About This Item

MDL number:
UNSPSC Code:
12352103
NACRES:
NA.22

$438.00


In StockDetails


Request a Bulk Order

General description

Totally effective surface-active cleaning agent for chemical, bacteriological and radiotechnical laboratory. Biodegradable.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. Should I dilute the product? How much? Thanks

    1 answer
    1. This product often is used as a 2% to 5% solution, where 20 mL to 50 mL of DCN 90 concentrate is diluted to 1000 mL with deionized water.

      Helpful?

  2. Is this product "30472 Sigma-Aldrich DCN 90" the same as "decon 90". Is this product from the same or different producent?

    1 answer
    1. The supplier for product 30472 is considered proprietary.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service