Skip to Content
Merck
All Photos(2)

Key Documents

Z273236

Sigma-Aldrich

CITRANOX® acid detergent

biodegradable

Synonym(s):

CITRANOX® acid detergent, biodegradable, biodegradable cleaner, low foaming acid detergent, metal oxide remover, phosphate-free cleaner, ultrasonic cleaner detergent

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$148.00
4 EA
$491.00

$148.00


Available to ship on28 April 2025Details



Select a Size

Change View
1 EA
$148.00
4 EA
$491.00

About This Item

UNSPSC Code:
47131800
NACRES:
NB.35

$148.00


Available to ship on28 April 2025Details


packaging

pkg of 1 × 3.8 L in 1EA
pkg of 4 × 3.8 L in 4EA

manufacturer/tradename

Alconox, Inc. 1801 (4 x 3.8 L)
Alconox, Inc. 1801-1 (3.8 L)

technique(s)

protein quantification: suitable

Looking for similar products? Visit Product Comparison Guide

General description

CITRANOX™ is a phosphate free, concentrated cleaner and metal brightener for high performance removal of metal oxides, scale, salts and inorganic residues in manual and ultrasonic cleaning. Corrosion inhibited and free rinsing for reliable results and no interfering residues. Ideal for trace metal analysis. USDA authorized. Dilute 2:100. pH 2.5.

Legal Information

CITRANOX is a registered trademark of Alconox, Inc.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service