Skip to Content
Merck
All Photos(1)

Documents

242985

Sigma-Aldrich

Alconox® detergent

bulk packed

Synonym(s):

anionic detergent, concentrated powdered detergent, general laboratory cleaner, lab detergent, low foaming detergent, ultrasonic cleaner detergent

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41103200
NACRES:
NB.35

Agency

suitable for ASTM® 7968
suitable for ASTM® 7979
suitable for EPA 1613
suitable for EPA 601
suitable for EPA 8327
suitable for SM 5210

packaging

box of 11.3 kg (25 lb)
pkg of drum of 45.4 kg (100 lb)

manufacturer/tradename

Alconox, Inc. 1101 (45.4 kg)
Alconox, Inc. 1125 (11.3 kg)

Looking for similar products? Visit Product Comparison Guide

General description

  • Alconox® detergent is a concentrated powdered cleaner for both manual and ultrasonic cleaning of glass, metal, and plastic.
  • It is used to clean various healthcare instruments, laboratory ware, vacuum equipment, tissue culture ware, personal protective equipment, etc.
  • Biodegradable and readily disposable - potential substitute for corrosive acids and hazardous solvents.
  • High performance, anionic and low foaming - does not damage the washer parts and leave behind any residue.
Powdered detergent for general lab cleaning.

Features and Benefits

  • Concentrated and long-lasting
  • Biodegradable and readily disposable
  • Penetrating wetting power to save
  • Alternative for corrosive acids and hazardous solvents
  • Easy to rinse, doesn’t form interfering residues, and provides reliable results

Legal Information

ALCONOX is a registered trademark of Alconox, Inc.
ASTM is a registered trademark of American Society for Testing and Materials

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Related Content

FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service