Skip to Content
Merck
All Photos(1)

Key Documents

627609

Sigma-Aldrich

Caspase Inhibitor I

InSolution

Synonym(s):

InSolution Caspase Inhibitor I

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$463.00

$463.00


Estimated to ship on28 May 2025



Select a Size

Change View
1 MG
$463.00

About This Item

Empirical Formula (Hill Notation):
C22H30FN3O7
Molecular Weight:
467.49
UNSPSC Code:
12352200
NACRES:
NA.54

$463.00


Estimated to ship on28 May 2025


Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

shipped in

wet ice

storage temp.

−20°C

General description

When using with a purified recombinant enzyme, pretreatment with an esterase is required.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Fas-mediated apoptosis in Jurkat cells and staurosporine-induced cell death in corneal epithelial cells
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 10 mM (1 mg in 214 µl) solution of Caspase Inhibitor I Cat. No. 627610 in DMSO.

Reconstitution

Following initial thaw, aliquot, purge with inert gas, and freeze(-20°C).

Analysis Note

Single spot by TLC

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - (refers to pure substance)

Flash Point(C)

87 °C - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service