Skip to Content
Merck
All Photos(2)

Key Documents

NIST984

Rubidium chloride

NIST SRM 984

Sign Into View Organizational & Contract Pricing

Select a Size

0.25 G
$1,300.00

$1,300.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
0.25 G
$1,300.00

About This Item

Linear Formula:
RbCl
CAS Number:
Molecular Weight:
120.92
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.24

$1,300.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

isotopic purity

 85Rb
 87Rb

packaging

pkg of 0.25 g

manufacturer/tradename

NIST®

technique(s)

mass spectrometry (MS): suitable

mp

715 °C (lit.)

density

2.8 g/mL at 25 °C (lit.)

format

neat

SMILES string

[Cl-].[Rb+]

InChI

1S/ClH.Rb/h1H;/q;+1/p-1

InChI key

FGDZQCVHDSGLHJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Rubidium chloride (RbCl)

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service