Skip to Content
Merck
All Photos(2)

Key Documents

NIST975A

Isotopic standard for chlorine

NIST® SRM® 975a

Synonym(s):

Sodium chloride, Halite, NaCl

Sign Into View Organizational & Contract Pricing

Select a Size

0.25 G
$1,390.00

$1,390.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
0.25 G
$1,390.00

About This Item

Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
Beilstein:
3534976
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.24

$1,390.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

isotopic purity

 35Cl
 37Cl

packaging

pkg of 0.25 g

manufacturer/tradename

NIST®

technique(s)

mass spectrometry (MS): suitable

mp

801 °C (lit.)

format

neat

SMILES string

[Na+].[Cl-]

InChI

1S/ClH.Na/h1H;/q;+1/p-1

InChI key

FAPWRFPIFSIZLT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

SRM 975a contains 0.25 g of sodium chloride (NaCl) with a purity of 99.89% by mass. For more information, please refer to the COA and SDS.

SRM 975A_cert

SRM 975A_SDS

Application

The Standard Reference Material (SRM) Isotopic standard for chlorine is intended for use as an isotopic standard.

Features and Benefits

  • Accompanied with certified values for the absolute abundance ratio of 35Cl/37Cl and isotopic compositions of 35Cl and 37Cl.
  • Maintenance and expiration of SRM certification, instructions for use are provided on NIST certificate.

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Chlorine-35 (Cl), Chlorine-37 (Cl)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service