Skip to Content
Merck
All Photos(1)

Key Documents

NIST3530

Iodized Table Salt (Iodide)

NIST®SRM®

Synonym(s):

Sodium chloride, Halite, NaCl

Sign Into View Organizational & Contract Pricing

Select a Size

200 G
$1,380.00

$1,380.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
200 G
$1,380.00

About This Item

Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
Beilstein:
3534976
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$1,380.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

packaging

pkg of 200 g

manufacturer/tradename

NIST®

mp

801 °C (lit.)

application(s)

food and beverages

format

matrix material

storage temp.

15-25°C

SMILES string

[Na+].[Cl-]

InChI

1S/ClH.Na/h1H;/q;+1/p-1

InChI key

FAPWRFPIFSIZLT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Each unit of SRM 3530 consists of a bottle containing ~200 g of commercially available iodized table salt, sealed inside an aluminized polyethylene terephthalate (PET) pouch, supplied with an oxygen absorber pack.

SRM 3530 _SDS

SRM3530 _CoA

Application

Iodized Table Salt standard reference material (SRM) is intended primarily for validation of methods used to determine the iodide concentration in an iodized table salt or similar material.

This SRM is also suitable for quality assurance when assigning values to in-house reference materials.

Features and Benefits

  • Available with a certificate documenting NIST-certified value of mass fraction of iodine as iodide. 
  • Assay value determined using gravimetric titrimetric, ICP-MS, and linear voltammetry methods. 
  • The NIST certificate is provided with storage and use instructions.

Other Notes

Certified for the analytes listed below.
Iodine (I)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service