Skip to Content
Merck
All Photos(1)

Key Documents

NIST2719

Calcined petroleum coke

NIST® SRM® 2719

Synonym(s):

Coke

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$1,380.00

$1,380.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 G
$1,380.00

About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

$1,380.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of 50 g

manufacturer/tradename

NIST®

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

Each unit of Standard reference material (SRM) Calcined petroleum coke contains 50 g calcined petroleum coke is ground to pass a 250 µm (60 mesh) sieve, homogenized, and then bottled under an argon atmosphere. For more information, please refer to the SDS and COA.

SRM 2719_cert

SRM 2719_SDS

Application

NIST2719is intended for calibrating apparatus and evaluating techniques used in theanalysis of calcined petroleum coke and other materials with a similar matrix.

Features and Benefits

  • NIST provides certified values for elements on a dry basis that are metrologically traceable to the SI-derived unit for massfraction, expressed as milligrams per kilogram.
  • Period of validity, use, storage, and instructions for drying are provided on the NIST certificate.

Other Notes

Certified for the analytes listed below.
Aluminum (Al), Calcium Ca), Carbon (C), Cobalt (Co), Hydrogen (H), Iron (Fe), Nickel (Ni), Nitrogen(N), Silicon(Si), Sodium(Na), Sulfur (S), Vanadium (V)

Reference/Informational value is provided for the following analytes.
Carbon (C), Cobalt (Co), Hydrogen (H), Nitrogen (N), Sodium (Na), Silicon (Si), Ash, Volatile Matter, Gross Calorific Value

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service