Skip to Content
Merck
All Photos(1)

Key Documents

NIST2781

Domestic sludge

NIST® SRM® 2781

Sign Into View Organizational & Contract Pricing

Select a Size

40 G
$1,770.00

$1,770.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
40 G
$1,770.00

About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

$1,770.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of 40 g

manufacturer/tradename

NIST®

application(s)

environmental

format

matrix material

General description

One unit of SRM 2781 consists of a bottle containing 40 g of dried and pulverized domestic sludge. For more information, please refer to the SDS and COA.
SRM 2781_cert
SRM 2781_SDS

Application

Domestic sludge Standard Reference Material (SRM) is intended primarily for the evaluation of methods employed in analyzing sludges and other materials of a similar matrix.

Features and Benefits

  • Accompanied with certified mass fraction values for elements that are metrologically traceable to the SI unit for mass expressed as percentages or milligrams per kilogram.
  • Reference mass fraction values are provided, along with both elemental and organic constituents, which are noncertified values serving as the best estimates of true values.
  • Information mass fraction values are also provided for organic constituents.
  • Expiration details, storage, and instructions for use are provided on the NIST certificate.

Other Notes

Certified for the analytes listed below.
Arsenic (As), Nitrogen (N), Cadmium (Cd), Chromium (Cr), Copper (Cu), Lead (Pb), Mercury (Hg), Molybdenum (Mo), Nickel (Ni), Selenium (Se), Silver (Ag), Zinc (Zn)

Reference/Informational value is provided for the following analytes.
Beryllium (Be), Aluminum (Al), Calcium (Ca), Iron (Fe), Magnesium (Mg), Phosphorus (P), Potassium (K), Silicon (Si), Sodium (Na), Titanium (Ti), 1,3,4,6,7,8-Hexahydro-4,6,6,7,8,8-hexa-methyl-cyclopenta-(γ)-2-benzopyran (HHCB), 7-Acetyl-1,1,3,4,4,6-hexamethyltetralin (AHTN), 5-Acetyl-1,1,2,6-tetramethyl-3-isopropylindan (ATII), 4-Acetyl-1,1-dimethyl-6-tert-butylindan (ADBI), 6-Acetyl-1,1,2,3,3,5-hexamethylindan (AHMI), Perfluorohexanoic Acid (PFHxA), Perfluoroheptanoic Acid (PFHpA), Perfluorooctanoic Acid (PFOA), Perfluorohexanesulfonic Acid (PFHxS), Perfluorooctanesulfonic Acid (PFOS), Perfluorooctane Sulfonamide (PFOSA), γ-hexabromocyclododecane (γ-HBCD), γ-hexabromocyclododecane (γ-HBCD), γ-hexabromocyclododecane (γ-HBCD), Aluminum (Al), Barium (Ba), Cadmium (Cd), Calcium (Ca), Chromium (Cr), Copper (Cu), Iron (Fe), Lead (Pb), Magnesium (Mg), Manganese (Mn), Nickel (Ni), Silver (Ag), Vanadium (V), Zinc (Zn), Perfluorononanoic Acid (PFNA), Perfluorodecanoic Acid (PFDA)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. From where was this fine sludge procured?

    1 answer
    1. The U.S. Geological Survey (USGS), under contract to NIST, obtained partially dehydrated sewage cake material from the Metropolitan Denver Sewage Disposal District No. 1. The material (approximately 182 kg) was placed in plastic-lined drums and transported to the USGS facilities (Lakewood, CO) for processing. It was dried at ambient temperature in a forced air chamber, ground to pass a 74 µm (200 mesh) sieve, blended for 24 h to assure homogeneity of the pulverized material. Test samples were taken from the blender for preliminary homogeneity analyses. The material was then radiation sterilized. The sterilized material was shipped in bulk to NIST, where the material was bottled in 40 g units after reblending for 4 h.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service