Skip to Content
Merck
All Photos(1)

Key Documents

NIST2775

Foundry coke

NIST® SRM® 2775

Synonym(s):

Coke

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$1,060.00

$1,060.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 G
$1,060.00

About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

$1,060.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of 50 g

manufacturer/tradename

NIST®

technique(s)

mass spectrometry (MS): suitable

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

This Standard Reference Material (SRM) is intended primarily for use in the evaluation of test methods and for the calibration of instruments used to determine sulfur in foundry (metallurgical) coke. Each unit of SRM 2775 consists of 50 g of foundry coke that was ground to pass a 60 mesh (250 μm) sieve, homogenized, and bottled under an argon atmosphere. For more information, plese refer to the SDS and COA.

SRM 2775_cert

SRM 2775_SDS

Other Notes

Certified for the analytes listed below.
Sulfur (S)

Reference/Informational value is provided for the following analytes.
Ash, Volatile Matter, Carbon (C), Hydrogen(H), Nitrogen(N)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service