Skip to Content
Merck
All Photos(1)

Key Documents

NIST1635A

Trace elements in coal (sub-bituminous)

NIST® SRM® 1635a

Synonym(s):

Coal powder

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$1,950.00

$1,950.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 G
$1,950.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$1,950.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of 50 g

manufacturer/tradename

NIST®

application(s)

environmental

format

matrix material

General description

A unit of Standard Reference Material (SRM) 1635a consists of 50 g of sub-bituminous coal that was ground to pass a 250μm (60 mesh) sieve, homogenized, bottled under an argon atmosphere, and sealed in an aluminized bag.

SRM 1635A_cert

SRM 1635A_SDS

Application

This SRM is intended primarily for the evaluation of techniques used in the analysis of coals and materials of a similar matrix.

Features and Benefits

  • Certificate of analysis lists the actual values of the components
  • Information on storage and handling is available in MSDS

Other Notes

Certified for the analytes listed below.
Barium (Ba), Sulfur (S), Iron (Fe), Strontium (Sr), Sodium (Na)
Trace elements : Antimony (Sb), Mercury (Hg), Arsenic (As), Nickel (Ni), Chromium (Cr), Rubidium (Rb), Cobalt (Co), Selenium (Se), Copper (Cu), Thorium (Th), Lead (Pb), Uranium (U), Manganese (Mn), Vanadium (V)

Reference/Informational value is provided for the following analytes.
Aluminum (Al), Calcium (Ca), Hydrogen (H), Magnesium (Mg), Potassium (K), Titanium (Ti), Boron (B), Hafnium (Hf), Cadmium (Cd), Molybdenum (Mo), Cerium (Ce), Samarium (Sm), Cesium (Cs), Scandium (Sc), Chlorine (Cl), Zinc (Zn), Europium (Eu), Bromine (Br)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service