Skip to Content
Merck
All Photos(1)

Key Documents

NIST2691

Coal fly ash

NIST® SRM® 2691

Sign Into View Organizational & Contract Pricing

Select a Size

3 X 10 G
$1,220.00

$1,220.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
3 X 10 G
$1,220.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$1,220.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of 3 x 10 g

manufacturer/tradename

NIST®

application(s)

environmental

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

SRM 2691 contains three hermetically sealed glass vials, each containing 10 g of fly ash pulverized to a particle size of less than 150 µm and thoroughly blended to ensure a high degree of homogeneity. For more information, please refer to the SDS and COA.
SRM 2691_cert
SRM 2691_SDS

Application

Coal fly ash standard Reference Material (SRM) is intended for use in assessing the analytical methods used to classify coal fly ash and to determine constituent elements in coal fly ash or similar matrix.

Features and Benefits

  • NIST Certified Mass Fraction values for constituent elements are traceable to NIST’s realization of the SI unit of the kilogram and expressed as a percentage.
  • Information values for constituent elements are expressed as mass fraction.
  • Expiration details and instructions for use, storage, and handling are provided on the NIST certificate.

Other Notes

Certified for the analytes listed below.
Aluminum (Al), Calcium (Ca), Iron (Fe), Magnesium (Mg), Phosphorus (P), Potassium (K), Silicon (Si), Sodium (Na), Sulfur (S), Titanium (Ti), Mercury (Hg)

Reference/Informational value is provided for the following analytes.
Antimony (Sb), Arsenic (As), Barium (Ba), Beryllium (Be), Cobalt (Co), Chromium (Cr), Cesium (Cs), Europium (Eu), Hafnium (Hf), Lead (Pb), Manganese (Mn), Nickel (Ni), Scandium (Sc), Selenium (Se), Strontium (Sr), Thorium (Th), Zinc (Zn), Aluminum Oxide (Al2O3), Barium oxide (BaO), Ferric Oxide (Fe2O3), Potassium oxide (K2O), Magnesium oxide (MgO), Manganese oxide (MnO), Sodium oxide (Na2O), Silicon dioxide (SiO2), Strontium oxide (SrO), Titanium dioxide (TiO2)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization and quantification of biochar alkalinity.
Fidel B R, et al.
Chemosphere, 167, 367-373 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service