Skip to Content
Merck
All Photos(1)

Key Documents

NIST1633C

Trace elements in coal fly ash

NIST® SRM® 1633c

Synonym(s):

Coal fly ash (ash; coal ash; fly ash; flyash; coal ash, by product; bottom ash)

Sign Into View Organizational & Contract Pricing

Select a Size

75 G
$1,860.00

$1,860.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
75 G
$1,860.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$1,860.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of 75 g

manufacturer/tradename

NIST®

application(s)

environmental

format

matrix material

General description

Standard Reference Material (SRM) is a bituminous coal fly ash that was sieved through a nominal sieve opening of 74μm (200 mesh) and then blended to assure homogeneity.

SRM 1633C_cert

SRM 1633C_SDS

Application

This SRM is intended for use in the evaluation of analytical methods for the determination of constituent elements in coal fly ash or materials of a similar matrix.

Features and Benefits

  • A unit of SRM 1633c consists of 75 g of powdered material
  • Details of composition and information on hazardous ingredients are provided in NIST certificate of analysis

Other Notes

Certified for the analytes listed below.
Aluminum (Al), Arsenic (As), Barium (Ba), Cadmium (Cd), Calcium (Ca), Cobalt (Co), Iron (Fe), Copper (Cu), Potassium (K), Mercury (Hg), Magnesium (Mg), Manganese (Mn), Sodium (Na), Nickel (Ni), Titanium (Ti), Lead (Pb), Rubidium (Rb), Antimony (Sb), Strontium (Sr), Vanadium (V)

Reference/Informational value is provided for the following analytes.
Sulfur (S), Chromium (Cr), Silicon (Si), Cesium (Cs), Phosphorus (P), Dysprosium (Dy), Europium (Eu), Lanthanum (La), Lutetium (Lu), Scandium (Sc), Selenium (Se), Tantalum (Ta), Terbium (Tb), Thorium (Th), Uranium (U), Zinc (Zn), Beryllium (Be), Cerium (Ce), Gallium (Ga), Hafnium (Hf), Indium (In), Neodymium (Nd), Samarium (Sm), Ytterbium (Yb)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service