79714
Sodium acetate
suitable for HPLC, LiChropur™, 99.0-101.0% (NT)
Synonym(s):
Acetic acid sodium salt
Select a Size
About This Item
Recommended Products
Quality Level
Assay
99.0-101.0% (NT)
form
solid
autoignition temp.
1112 °F
quality
LiChropur™
technique(s)
HPLC: suitable
pH
8.5-10.5
pKa
4.76 (acetic acid)
mp
>300 °C (dec.) (lit.)
solubility
H2O: 10%, clear, colorless
UV absorption
λ: 250 nm Amax: ≤0.05
λ: 260 nm Amax: ≤0.03
λ: 280 nm Amax: ≤0.02
λ: 300 nm Amax: ≤0.01
λ: 500 nm Amax: ≤0.01
SMILES string
[Na+].CC([O-])=O
InChI
1S/C2H4O2.Na/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1
InChI key
VMHLLURERBWHNL-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Application
- Fast automated online xylanase activity assay using HPAEC-PAD.: This study utilizes sodium acetate in a novel automated method for xylanase activity measurement, enhancing analytical efficiency and precision (Cürten et al., 2018).
- Preparation and properties of type-specific M antigen isolated from a group A, type 1 hemolytic streptococcus.: The study uses sodium acetate for preparing bacterial antigens, showing its utility in microbiological and immunological analyses (Lancefield and Perlmann, 1952).
Legal Information
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service