5.43837
Ammonium dihydrogen phosphate
for HPLC LiChropur™
Synonym(s):
Ammonium dihydrogen phosphate, Ammonium biphosphate, Ammonium phosphate monobasic, Primary ammonium phosphate, Monoammonium orthophosphate
About This Item
Recommended Products
vapor pressure
0.066 hPa ( 125 °C)
Quality Level
form
solid
technique(s)
HPLC: suitable
pH
3.8-4.4 (20 °C, 50 g/L in H2O)
mp
190 °C
solubility
368 g/L
density
1.80 g/cm3 at 20 °C
bulk density
800‑1100 kg/m3
storage temp.
2-30°C
SMILES string
[P](=O)([O-])([O-])[O-].[N+H4].[H+].[H+]
InChI
1S/H3N.H3O4P/c;1-5(2,3)4/h1H3;(H3,1,2,3,4)
InChI key
LFVGISIMTYGQHF-UHFFFAOYSA-N
Related Categories
Application
- Synthesis and characterization of hydroxyapatite nanoparticles from calcium hydroxide fouled with gases evolved from smokestack of glass industry.: This research explores the synthesis of hydroxyapatite nanoparticles using ammonium dihydrogen phosphate, detailing the process and analytical chemistry techniques involved. The study provides insights into the potential environmental and industrial applications of this method, offering an innovative approach to materials synthesis (Abdelmoaty and Mousa, 2024).
- Acid etching-induced nanocutting of LaNiO(3) transplanting self-assembled photodiode array-like LaNiO(3)/N,P-RGO nanoreactor for efficient photocatalytic hydrogen evolution.: This article discusses the use of ammonium dihydrogen phosphate in creating LaNiO(3) based nanoreactors through acid etching techniques. The resulting materials show promising applications in photocatalysis and hydrogen production, illustrating the compound′s utility in renewable energy technologies (Zou et al., 2024).
- Sustainable and Environmentally Friendly Microwave Synthesis of Nano-Hydroxyapatite from Decarbonized Eggshells.: This study utilizes ammonium dihydrogen phosphate in a green chemistry process to synthesize nano-hydroxyapatite from eggshells, emphasizing the environmental benefits and sustainability of this approach in materials science (Mahmoud, 2024).
- A Self-Powered Biochemical Sensor for Intelligent Agriculture Enabled by Signal Enhanced Triboelectric Nanogenerator.: This innovative research uses ammonium dihydrogen phosphate in the development of a self-powered biochemical sensor for agricultural applications, highlighting its role in enhancing device sensitivity and sustainability (Gao et al., 2024).
- Therapeutic drug monitoring of vancomycin in surgical patients through a validated HPLC method.: In this clinical study, ammonium dihydrogen phosphate is employed as part of a high-performance liquid chromatography (HPLC) method for monitoring vancomycin in surgical patients, demonstrating its crucial role in improving analytical accuracy and patient care (Akram et al., 2024).
Legal Information
related product
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Learn how the Purospher® STAR RP-18e Hibar® HPLC column meets the system suitability criteria outlined in the USP monograph for the fluticasone propionate assay.
LC-MS/MS quantitative method analyzes highly polar 11 nucleotide activated sugars using Supel™ Carbon LC column.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service