Skip to Content
Merck
All Photos(1)

Key Documents

7510-OP

Millipore

Sodium Acetate

OmniPur® Grade, >=99.0%, Anhydrous

Synonym(s):

OmniPur® Sodium Acetate, Anhydrous

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$86.80

$86.80


Estimated to ship on26 March 2025


Request a Bulk Order

Select a Size

Change View
500 G
$86.80

About This Item

Empirical Formula (Hill Notation):
C2H3NaO2
CAS Number:
Molecular Weight:
82.03
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.21

$86.80


Estimated to ship on26 March 2025


Request a Bulk Order

Quality Level

form

crystals

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

shipped in

ambient

SMILES string

[Na+].[O-]C(=O)C

InChI

1S/C2H4O2.Na/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1

InChI key

VMHLLURERBWHNL-UHFFFAOYSA-M

General description

Molecular biology grade.

Specifications:
A (260 nm, 0.1 M, water): 0.01AU max
A (280 nm, 0.1 M, water): 0.01AU max
Assay: 99.0% min
DNase: None detected
Heavy metals (as Pb): 0.001% max
pH (5%, water): 7.5-9.2
Phosphatase: None detected
Protease: None detected
RNase: None detected





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service